missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxytoluene monohydrate, 97%, Thermo Scientific Chemicals

Product Code. 11468653
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-39-5
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00149092
InChI Key NBKPNAMTHBIMLA-UHFFFAOYSA-N
Synonym 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate
PubChem CID 3083941
IUPAC Name 5-methylbenzene-1,3-diol;hydrate
SMILES CC1=CC(=CC(=C1)O)O.O

Specifications

Melting Point 56°C to 60°C
Boiling Point 147°C (5 mmHg)
Quantity 5 g
Beilstein 5103474
Sensitivity Air Sensitive
Merck Index 14,6864
Formula Weight 142.16 (124.14 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3,5-Dihydroxytoluene monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'