Learn More
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals
34.57 € - 264.32 €
Chemical Identifiers
CAS | 2478-38-8 |
---|---|
Molecular Formula | C10H12O4 |
Molecular Weight (g/mol) | 196.20 |
MDL Number | MFCD00008748 |
InChI Key | OJOBTAOGJIWAGB-UHFFFAOYSA-N |
Synonym | acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy |
PubChem CID | 17198 |
ChEBI | CHEBI:2404 |
IUPAC Name | 1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone |
SMILES | COC1=CC(=CC(OC)=C1O)C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10102910
|
Thermo Scientific Acros
115540010 |
1 g | Glass bottle |
34.57 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10346130
|
Thermo Scientific
115540050 |
5 g | Glass bottle |
77.79 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10339273
|
Thermo Scientific Acros
115540250 |
25 g | Glass bottle |
264.32 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2478-38-8 | |
196.20 | |
OJOBTAOGJIWAGB-UHFFFAOYSA-N | |
17198 | |
1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone |
C10H12O4 | |
MFCD00008748 | |
acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy | |
CHEBI:2404 | |
COC1=CC(=CC(OC)=C1O)C(C)=O |
Specifications
2478-38-8 | |
100.0 | |
Beige to Brown | |
97% | |
HOC6H2(OCH3)2COCH3 | |
08,II,444 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,acetone,ether | |
COC1=CC(=CC(OC)=C1O)C(C)=O | |
196.20 | |
CHEBI:2404 | |
97% | |
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97% |
96.0 | |
123°C to 128°C | |
Authentic | |
C10H12O4 | |
MFCD00008748 | |
acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxy | |
OJOBTAOGJIWAGB-UHFFFAOYSA-N | |
1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone | |
17198 | |
196.2 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 219-610-5
missing translation for 'rtecsNumber' : AM8440000
missing translation for 'tsca' : TSCA
- 3',5'-Dimethoxy-4'-hydroxyacetophenone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only