Learn More
3',5'-Dimethoxyacetophenone, 97%, Thermo Scientific Chemicals
76.10 € - 849.00 €
Chemical Identifiers
CAS | 39151-19-4 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.20 |
MDL Number | MFCD00008739 |
InChI Key | YJKHOUIVWKQRSL-UHFFFAOYSA-N |
Synonym | 3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889 |
PubChem CID | 95997 |
IUPAC Name | 1-(3,5-dimethoxyphenyl)ethanone |
SMILES | COC1=CC(=CC(OC)=C1)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430738
|
Thermo Scientific Alfa Aesar
B25224.03 |
1 g |
76.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440738
|
Thermo Scientific Alfa Aesar
B25224.06 |
5 g |
229.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450738
|
Thermo Scientific Alfa Aesar
B25224.14 |
25 g | N/A | ||||||
Description
3',5'-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3′,5′-Dimethoxyacetophenone is used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole1. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.
Solubility
Insoluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
39151-19-4 | |
180.20 | |
YJKHOUIVWKQRSL-UHFFFAOYSA-N | |
95997 | |
COC1=CC(=CC(OC)=C1)C(C)=O |
C10H12O3 | |
MFCD00008739 | |
3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889 | |
1-(3,5-dimethoxyphenyl)ethanone |
Specifications
39151-19-4 | |
290°C to 291°C | |
C10H12O3 | |
MFCD00008739 | |
3',5'-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethanone, 3,5-dimethoxyacetophenone, 1-3,5-dimethoxyphenyl ethan-1-one, ethanone, 1-3,5-dimethoxyphenyl, 1-acetyl-3,5-dimethoxybenzene, acetophenone, 3',5'-dimethoxy-6ci,7ci, acmc-20anci, pubchem3391, opera_id_1889 | |
YJKHOUIVWKQRSL-UHFFFAOYSA-N | |
1-(3,5-dimethoxyphenyl)ethanone | |
95997 | |
97% |
39°C to 41°C | |
>110°C (230°F) | |
1.543 | |
2045586 | |
Insoluble in water. | |
COC1=CC(=CC(OC)=C1)C(C)=O | |
180.20 | |
180.2 | |
3',5'-Dimethoxyacetophenone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 254-322-3
missing translation for 'rtecsNumber' : KM5775717
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.