missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11423744
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylhydrazine hydrochloride participates inFischer indole synthesis protocol in biological evaluation of novel antitubercular indolecarboxamide derivatives against drug-susceptible and drug-resistant mycobacterium tuberculosis strains.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60481-36-9
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00052269
InChI Key RSBQANBNDXZFIY-UHFFFAOYSA-N
Synonym 3,5-dimethylphenylhydrazine hydrochloride, 3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenylhydrazinehydrochloride, 3,5-dimethyl phenylhydrazine hydrochloride, 3,5-dimethylphenylhydrazine, hydrazine, 3,5-dimethylphenyl-, monohydrochloride, pubchem6407, ksc491e7r, 1-3,5-dimethylphenyl hydrazine hydrochloride, 3,5-dimethylphenyl hydrazine-hydrogen chloride 1/1
PubChem CID 12651341
IUPAC Name (3,5-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=CC(=CC(=C1)NN)C.Cl

Specifications

Melting Point 180°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylphenylhydrazine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.