missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline, 97%, Thermo Scientific™

Product Code. 10516344
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10185-69-0
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD08443707
InChI Key CTRGRIHPFAVSOF-UHFFFAOYSA-N
Synonym 3-5-methyl-1,2,4-oxadiazol-3-yl aniline, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylamine, benzenamine,3-5-methyl-1,2,4-oxadiazol-3-yl
PubChem CID 11423869
IUPAC Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
SMILES CC1=NC(=NO1)C1=CC(N)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.