missing translation for 'onlineSavingsMsg'
Learn More
3,6-Di-tert-butylfluorene, 98%, Thermo Scientific Chemicals Product Code.: 11486886

3,6-Di-tert-butylfluorene, 98%, Thermo Scientific Chemicals

Product Code. 11486886
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11486886

Brand: Thermo Scientific Alfa Aesar B23996.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58775-07-8
Molecular Formula C21H26
Molecular Weight (g/mol) 278.44
MDL Number MFCD02683109
InChI Key IWUHFBNEZZKUEY-UHFFFAOYSA-N
PubChem CID 11140437
SMILES CC(C)(C)C1=CC2=C(CC3=C2C=C(C=C3)C(C)(C)C)C=C1

Specifications

Melting Point 79°C to 82°C
Quantity 5 g
Synonym 3,6-di-tert-butylfluorene, 3,6-di-tert-butyl-9h-fluorene, acmc-1atta, 3,6-di-t-butylfluorene, 3,6-di-tert-butyl-fluorene, 3,6-bis tert-butyl fluorene
IUPAC Name 3,6-di-tert-butyl-9H-fluorene
Formula Weight 278.44
Percent Purity 98%
Chemical Name or Material 3,6-Di-tert-butylfluorene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246148 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberB23996.03,B23996.06
Lot Number10243343 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberB23996.03,B23996.06
Lot Number10243197 Certificate TypeCertificate of Analysis Date29/08/2022 Catalog NumberB23996.03,B23996.06
Lot Number10203831 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23996.03,B23996.06
Lot Number10148529 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23996.03,B23996.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,6-Di-tert-butylfluorene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.