missing translation for 'onlineSavingsMsg'
Learn More

3',6'-Dichloro-2'-fluoroacetophenone, 97%, Thermo Scientific™

Product Code. 15454837
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-72-9
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.025
MDL Number MFCD09025392
InChI Key KIPVBHCZTSPODV-UHFFFAOYSA-N
Synonym 3',6'-dichloro-2'-fluoroacetophenone, 1-3,6-dichloro-2-fluorophenyl ethanone, ethanone, 1-3,6-dichloro-2-fluorophenyl, 1-acetyl-3,6-dichloro-2-fluorobenzene
PubChem CID 46737559
IUPAC Name 1-(3,6-dichloro-2-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1F)Cl)Cl

Specifications

Refractive Index 1.5325
Quantity 250 mg
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 3',6'-Dichloro-2'-fluoroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'