missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Product Code. 11975761
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181180-42-7
Molecular Formula C6H7F3O3S2
Molecular Weight (g/mol) 248.24
InChI Key ZMDCEXLCDOPKGH-UHFFFAOYSA-N
Synonym 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 2,3dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 3,6-dihydro-2h-thiopyran-4-yltrifluoromethanesulfonate, trifluoro-methanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, methanesulfonic acid, trifluoro-, 3,6-dihydro-2h-thiopyran-4-yl ester
PubChem CID 22625751
IUPAC Name 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
SMILES C1CSCC=C1OS(=O)(=O)C(F)(F)F

Specifications

Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 248.24
Percent Purity 97%
Chemical Name or Material 3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
compliance-icons compliance-icons
Product Identifier
  • 3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.