missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals
48.04 € - 161.81 €
Chemical Identifiers
CAS | 40932-63-6 |
---|---|
Molecular Formula | C13H12OS |
Molecular Weight (g/mol) | 216.30 |
MDL Number | MFCD00151790 |
InChI Key | GREAZYFTAJMZFD-UHFFFAOYSA-N |
Synonym | 3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone |
PubChem CID | 2728765 |
IUPAC Name | 1-(2-methyl-5-phenylthiophen-3-yl)ethanone |
SMILES | CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303147
|
Thermo Scientific Alfa Aesar
L13554.03 |
1 g | N/A | ||||||
11313147
|
Thermo Scientific Alfa Aesar
L13554.06 |
5 g |
161.81 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40932-63-6 | |
216.30 | |
GREAZYFTAJMZFD-UHFFFAOYSA-N | |
2728765 | |
CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1 |
C13H12OS | |
MFCD00151790 | |
3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone | |
1-(2-methyl-5-phenylthiophen-3-yl)ethanone |
Specifications
40932-63-6 | |
C13H12OS | |
8481276 | |
GREAZYFTAJMZFD-UHFFFAOYSA-N | |
1-(2-methyl-5-phenylthiophen-3-yl)ethanone | |
2728765 | |
98% |
67°C to 69°C | |
MFCD00151790 | |
3-acetyl-2-methyl-5-phenylthiophene, 1-2-methyl-5-phenylthiophen-3-yl ethanone, 1-2-methyl-5-phenylthien-3-yl ethanone, ethanone, 1-2-methyl-5-phenyl-3-thienyl, acmc-20anao, 3-acetyl-2-methyl-5-phenylthiohene, 1-2-methyl-5-phenyl-3-thienyl ethanone, 1-2-methyl-5-phenyl-3-thiophenyl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl ethanone, 1-2-methyl-5-phenyl-thiophen-3-yl-ethanone | |
CC(=O)C1=C(C)SC(=C1)C1=CC=CC=C1 | |
216.30 | |
216.3 | |
3-Acetyl-2-methyl-5-phenylthiophene |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title