missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals

66.50 € - 255.00 €

Chemical Identifiers

CAS 139042-59-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD04974527
InChI Key MUKKGHQBUKOMTD-UHFFFAOYSA-N
Synonym 5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij
PubChem CID 15668195
IUPAC Name 1-(6-bromopyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H33204.03
1 g
66.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
15475997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H33204.06
5 g
255.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

139042-59-4
200.035
MUKKGHQBUKOMTD-UHFFFAOYSA-N
15668195
CC(=O)C1=CN=C(C=C1)Br
C7H6BrNO
MFCD04974527
5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij
1-(6-bromopyridin-3-yl)ethanone

Specifications

139042-59-4
C7H6BrNO
5-acetyl-2-bromopyridine, 1-6-bromopyridin-3-yl ethanone, 2-bromo-5-acetylpyridine, 1-6-bromo-pyridin-3-yl-ethanone, 1-6-bromopyridin-3-yl ethan-1-one, 3-acetyl-6-bromopyridine, ethanone, 1-6-bromo-3-pyridinyl, 1-6-bromopyrid-3-yl ethanone, 1-6-bromo-3-pyridyl ethanone, acmc-209cij
CC(=O)C1=CN=C(C=C1)Br
200.035
200.03
3-Acetyl-6-bromopyridine
124°C to 128°C
MFCD04974527
MUKKGHQBUKOMTD-UHFFFAOYSA-N
1-(6-bromopyridin-3-yl)ethanone
15668195
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only