missing translation for 'onlineSavingsMsg'
Learn More

3-Acetylphenanthrene, 97%, Thermo Scientific Chemicals

Product Code. 11954841
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-76-1
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.27
InChI Key JKVNPRNAHRHQDD-UHFFFAOYSA-N
Synonym 3-acetylphenanthrene, 1-phenanthren-3-yl ethanone, ethanone, 1-3-phenanthrenyl, 1-3-phenanthryl ethanone, methyl 3-phenanthryl ketone, ketone, methyl 3-phenanthryl, 1-phenanthren-3-yl ethan-1-one, acmc-1clh9, dsstox_cid_29099, dsstox_rid_83318
PubChem CID 74867
IUPAC Name 1-phenanthren-3-ylethanone
SMILES CC(=O)C1=CC2=C(C=CC3=CC=CC=C32)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73.0°C to 76.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 25 g
Formula Weight 220.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Acetylphenanthrene
Show More Show Less
compliance-icons
Product Identifier
  • 3-Acetylphenanthrene, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'