missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetylpyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
Solubility
Soluble in Chloroform. Miscible with water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.
Solubility
Soluble in Chloroform. Miscible with water.
Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 12°C to 13°C |
Density | 1.106 |
Boiling Point | 218°C to 220°C |
Flash Point | 104°C (219°F) |
Odor | Characteristic |
Refractive Index | 1.535 |
Quantity | 100 g |
UN Number | UN2810 |
Beilstein | 107751 |
Merck Index | 14,6116 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'