missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1H-1,2,4-triazole, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
Specifications
Specifications
Melting Point | 147°C to 153°C |
Density | 1.138 |
Assay Percent Range | 96% |
Quantity | 250 g |
UN Number | UN3077 |
Beilstein | 107687 |
Merck Index | 14,489 |
Solubility Information | It is soluble in water,methanol,ethanol and chloroform,and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone. |
Formula Weight | 84.08 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- 3-Amino-1,2,4-triazole
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Amitrole
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'