missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-3-(2-nitrophenyl)propionic acid, 98%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
Specifications
Specifications
| Solubility Information | Slightly soluble in water. |
| Quantity | 5 g |
| Formula Weight | 210.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Amino-3-(2-nitrophenyl)propionic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction