missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorophenol, 97%, Thermo Scientific Chemicals

Product Code. 15422188
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-chlorophenol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic chemical intermediate for synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16026-77-0
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD03427127
InChI Key FWUALUHYKLDYAN-UHFFFAOYSA-N
PubChem CID 5070078
IUPAC Name 3-amino-4-chlorophenol
SMILES C1=CC(=C(C=C1O)N)Cl

Specifications

Melting Point 151°C to 154°C
Color Brown to Orange
Quantity 25 g
Synonym 4-chloro-3-aminophenol, acmc-1bo5g, 2-chloro-5-hydroxyaniline, phenol,3-amino-4-chloro, 3-azanyl-4-chloranyl-phenol, phenol, 3-amino-4-chloro, 3-amino-4-chlorophenol
Solubility Information Slightly soluble in water.
Formula Weight 143.57
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-4-chlorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.