missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11956121
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 873566-75-7
Melting Point 226°C to 232°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H7BFNO2
MDL Number MFCD03092925
Quantity 1 g
Synonym 3-amino-4-fluorophenyl boronic acid, 3-amino-4-fluorobenzeneboronic acid, 5-borono-2-fluoroaniline, 3-amino-4-fluoro-phenyl boronic acid, boronic acid, 3-amino-4-fluorophenyl, boronic acid,b-3-amino-4-fluorophenyl, hydrochloride, pubchem1743, acmc-209qjr, 3-amino-4-fluoro phenylboronic acid
InChI Key SYBMNJPUZMUPGQ-UHFFFAOYSA-N
SMILES NC1=C(F)C=CC(=C1)B(O)O
IUPAC Name (3-amino-4-fluorophenyl)boronic acid
Molecular Weight (g/mol) 154.94
PubChem CID 21954814
Formula Weight 154.94
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Amino-4-fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'