missing translation for 'onlineSavingsMsg'
Learn More

3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, Thermo Scientific™

Product Code. 10566015
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-60-1
Molecular Formula C6H8N2O7S
Molecular Weight (g/mol) 252.197
InChI Key HJASIPICABEOAV-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxy-5-nitrobenzenesulfonic acid hydrate, 3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate, benzenesulfonic acid,3-amino-4-hydroxy-5-nitro-, hydrate 1:1, benzenesulfonic acid,3-amino-4-hydroxy-5-nitro-,hydrate 1:1, 3-amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid-water 1/1
PubChem CID 5712259
IUPAC Name 3-amino-4-hydroxy-5-nitrobenzenesulfonic acid;hydrate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)N)S(=O)(=O)O.O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 252.21
Percent Purity 97%
Chemical Name or Material 3-Amino-4-hydroxy-5-nitrobenzene-1-sulfonic acid hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.