missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-hydroxybenzoic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 10146433
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14206-69-0
Molecular Formula C7H8ClNO3
Molecular Weight (g/mol) 189.60
MDL Number MFCD00043420
InChI Key CXESTILCPSBCGQ-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxybenzoic acid hydrochloride, ahba hydrochloride, 3-amino-5-hydroxybenzoic acid hcl, 3-amino-5-hydroxy-benzoic acid hydrochloride, benzoic acid, 3-amino-5-hydroxy-, hydrochloride, 3-amino-5-carboxyphenol hydrochloride, 3-carboxy-5-hydroxyanilne hydrochloride, 3-amino-5-hydroxybenzoic acid, chloride, 3-amino-5-hydroxybenzoicacidhydrochloride, 3-amino-5-hydroxybenzoic acid-hydrogen chloride 1/1
PubChem CID 24229779
IUPAC Name 3-amino-5-hydroxybenzoic acid;hydrochloride
SMILES Cl.NC1=CC(O)=CC(=C1)C(O)=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.62
Percent Purity 95%
Chemical Name or Material 3-amino-5-hydroxybenzoic acid hydrochloride hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'