missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-hydroxybenzoic acid hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 207.62 |
Percent Purity | 95% |
Chemical Name or Material | 3-amino-5-hydroxybenzoic acid hydrochloride hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'