missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-9-ethylcarbazole, 95%, Thermo Scientific Chemicals

Product Code. 11379769
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Peroxidase substrate for immunoblotting and immunohistochemical staining procedures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-32-1
Molecular Formula C14H14N2
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004964
InChI Key OXEUETBFKVCRNP-UHFFFAOYSA-N
Synonym 3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine
PubChem CID 8588
IUPAC Name 9-ethylcarbazol-3-amine
SMILES CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31

Specifications

Melting Point 98°C to 100°C
Odor Odorless
Quantity 25 g
Formula Weight 210.28
Percent Purity 95%
Chemical Name or Material 3-Amino-9-ethylcarbazole
compliance-icons compliance-icons
Product Identifier
  • 3-Amino-9-ethylcarbazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
Hazard Statement
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-Amino-9-ethylcarbazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.