missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzene-1-sulfonamide, 97%, Thermo Scientific™

Product Code. 10784013
Click to view available options
Quantity:
1 g
25 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 98-18-0
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key JPVKCHIPRSQDKL-UHFFFAOYSA-N
Synonym metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide
PubChem CID 7377
IUPAC Name 3-aminobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula NH2C6H4SO2NH2
Quantity 1 g
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material 3-Aminobenzene-1-sulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'