missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid, 98%, Thermo Scientific Chemicals

39.20 € - 560.79 €

Chemical Identifiers

CAS 30418-59-8
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD00007755
InChI Key JMZFEHDNIAQMNB-UHFFFAOYSA-N
Synonym 3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acid
PubChem CID 92269
IUPAC Name (3-aminophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)N)(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15409687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52909.03
1 g
39.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
15419687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52909.06
5 g
141.71 €
5g
Please to purchase this item. Need a web account? Register with us today!
15429687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52909.14
25 g
560.79 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid is used as an organic chemical synthesis intermediate. It is a boronic acid with potential use for biochemical research.

Solubility
Soluble in water (partly miscible).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents and heat.
Specifications

Chemical Identifiers

30418-59-8
136.945
JMZFEHDNIAQMNB-UHFFFAOYSA-N
92269
B(C1=CC(=CC=C1)N)(O)O
C6H8BNO2
MFCD00007755
3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acid
(3-aminophenyl)boronic acid

Specifications

30418-59-8
C6H8BNO2
3-aminobenzeneboronic acid, 3-aminophenyl boronic acid, m-aminophenylboronic acid, 3-aminophenyl boranediol, m-aminophenyl boronic acid, 3-amino phenylboronic acid, chembl20852, boronic acid, 3-aminophenyl, m-aminophenyl metaboric acid
JMZFEHDNIAQMNB-UHFFFAOYSA-N
(3-aminophenyl)boronic acid
92269
98%
138°C to 143°C
MFCD00007755
Soluble in water (partly miscible).
B(C1=CC(=CC=C1)N)(O)O
136.945
136.95
3-Aminobenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 250-189-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • 3-Aminobenzeneboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : (m-Aminophenyl)metaboric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only