missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid monohydrate, 97%, Thermo Scientific Chemicals

Product Code. 11493707
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.

Solubility
Soluble in DMSO and methanol.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206658-89-1
Molecular Formula C6H10BNO3
Molecular Weight (g/mol) 154.96
MDL Number MFCD00149554
InChI Key XAEOVQODHLLNKX-UHFFFAOYSA-N
Synonym 3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate, boronic acid, 3-aminophenyl-, monohydrate, 3-aminobenzeneboronic acid hydrate, pubchem4016, acmc-1cfgg, 3-boronoaniline monohydrate
PubChem CID 14389423
IUPAC Name (3-aminophenyl)boronic acid;hydrate
SMILES O.NC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 93°C to 97°C
Quantity 1 g
Beilstein 2936342
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO and methanol.
Formula Weight 154.96 (136.95 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Aminobenzeneboronic acid monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'