missing translation for 'onlineSavingsMsg'
Learn More
3-Aminoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals Product Code.: 15424948

3-Aminoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15424948
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15424948

Brand: Thermo Scientific Alfa Aesar H58726.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an intermediate in the production of mutagenic pyrolyzates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in the production of mutagenic pyrolyzates.

Solubility
Slightly Soluble in water. (4.9 g/L) (25°C)

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28036-33-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD06660421
InChI Key RTOFMMILZZGGNS-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-3-amine, 3-aminoimidazo 1,2-a pyridine, 3-amino-imidazo 1,2-a pyridine, acmc-20an9p, imidazopyridinamine, imidazo 1,2-a pyridin-3-ylamine, 4-hydroimidazo 1,2-a pyridine-3-ylamineShow MoreShow Less
PubChem CID 432221
IUPAC Name imidazo[1,2-a]pyridin-3-amine
SMILES C1=CC2=NC=C(N2C=C1)N

Specifications

Melting Point 114°C to 126°C
Quantity 1 g
Solubility Information Slightly Soluble in water. (4.9g/L) (25°C)
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 3-Aminoimidazo[1,2-a]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW08A027 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberH58726.03
Lot NumberU07A023 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58726.06,H58726.03
Lot NumberW08A027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58726.06,H58726.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminoimidazo[1,2-a]pyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.