missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-AminomethylAlizarin-N,N-Diacetic Acid, Pure, Indicator Grade, Fisher Chemical
Click to view available options
1g, Glass Jar
Specifications
Specifications
CAS | 3952-78-1 |
Melting Point | 190°C |
Color | Orange to Yellow |
Packaging | Glass Jar |
Molecular Formula | C19H15NO8 |
MDL Number | 149067 |
Quantity | 1 g |
Formula Weight | 421.35g/mol |
Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction