missing translation for 'onlineSavingsMsg'
Learn More
3-Aminooxetane, 95%, Thermo Scientific Chemicals Product Code.: 15493768

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Product Code. 15493768
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15493768

Brand: Thermo Scientific Alfa Aesar H57248.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine.

Solubility
Slightly soluble in water

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21635-88-1
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.095
MDL Number MFCD08544363
InChI Key OJEOJUQOECNDND-UHFFFAOYSA-N
Synonym 3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgamShow MoreShow Less
PubChem CID 9833923
IUPAC Name oxetan-3-amine
SMILES C1C(CO1)N

Specifications

Color Colorless to Yellow
Density 1.05 g/mL
Boiling Point 99°C
Flash Point 24°C (75°F)
Quantity 500 mg
UN Number UN1993
Sensitivity Air sensitive
Solubility Information Slightly soluble in water
Formula Weight 73.09
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminooxetane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16F068 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57248.MD,H57248.ME
Lot NumberC04Z010 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57248.MD,H57248.ME
Lot NumberG25Y007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57248.MD,H57248.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminooxetane, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.