missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopentane, 98+%, Thermo Scientific Chemicals

29.45 € - 259.29 €

Chemical Identifiers

CAS 616-24-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008096
InChI Key PQPFFKCJENSZKL-UHFFFAOYSA-N
Synonym 3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
PubChem CID 12019
ChEBI CHEBI:84248
IUPAC Name pentan-3-amine
SMILES CCC(N)CC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10753065
View Documents
Thermo Scientific Acros
183700050
5 g Glass bottle
29.45 €
5g
Please to purchase this item. Need a web account? Register with us today!
10360594
View Documents
Thermo Scientific Acros
183700250
25 g Glass bottle
91.05 €
25g
Please to purchase this item. Need a web account? Register with us today!
10320644
View Documents
Thermo Scientific Acros
183701000
100 g Glass bottle
259.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-24-0
87.17
PQPFFKCJENSZKL-UHFFFAOYSA-N
12019
pentan-3-amine
C5H13N
MFCD00008096
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
CHEBI:84248
CCC(N)CC

Specifications

616-24-0
100.0
0.7400g/mL
2°C
98% min. (GC)
(C2H5)2CHNH2
MFCD00008096
0.74
Solubility in water: soluble.
CCC(N)CC
87.17
CHEBI:84248
98+%
3-Aminopentane
98.0
Colorless to Yellow
89.0°C to 91.0°C
Authentic
C5H13N
1.4055
04, 179
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
PQPFFKCJENSZKL-UHFFFAOYSA-N
pentan-3-amine
12019
87.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-471-6

Product Identifier
  • 3-Aminopentane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only