Learn More
3-Aminophenol, 99%, Thermo Scientific Chemicals
25.73 € - 148.74 €
Chemical Identifiers
CAS | 591-27-5 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.13 |
MDL Number | MFCD00007786 |
InChI Key | CWLKGDAVCFYWJK-UHFFFAOYSA-N |
Synonym | m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg |
PubChem CID | 11568 |
ChEBI | CHEBI:28924 |
IUPAC Name | 3-aminophenol |
SMILES | NC1=CC=CC(O)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10249263
|
Thermo Scientific Acros
146100050 |
5 g | Glass bottle |
25.73 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386590
|
Thermo Scientific Acros
146102500 |
250 g | Glass bottle |
53.67 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10319323
|
Thermo Scientific Acros
146100010 |
1 kg | Glass bottle |
148.74 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-27-5 | |
109.13 | |
CWLKGDAVCFYWJK-UHFFFAOYSA-N | |
11568 | |
3-aminophenol |
C6H7NO | |
MFCD00007786 | |
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg | |
CHEBI:28924 | |
NC1=CC=CC(O)=C1 |
Specifications
591-27-5 | |
100.0 | |
Gray or Pink to White | |
155°C | |
98.5% min. (GC) | |
H2NC6H4OH | |
13, 401 | |
m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg | |
CWLKGDAVCFYWJK-UHFFFAOYSA-N | |
3-aminophenol | |
11568 | |
109.13 | |
Crystalline Powder |
98.5 | |
121.0°C to 124.0°C | |
164°C (15.0 hPa) | |
Authentic | |
C6H7NO | |
MFCD00007786 | |
15, 456 | |
Solubility in water: 35g/L (20°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol, ether and amyl alcohol, slightly soluble in benzene, very slightly soluble in petroleum ether | |
NC1=CC=CC(O)=C1 | |
109.13 | |
CHEBI:28924 | |
99% | |
3-Aminophenol, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-711-2
missing translation for 'rtecsNumber' : SJ4900000
missing translation for 'tsca' : TSCA
- 3-Aminophenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 3-Aminophenol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.