missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11412028
Click to view available options
Quantity:
5 g
25 g
100 g

Alfa Aesar 3-Aminophthalhydrazide, 98%, Quantity: 25g, Melting Point: 325 deg.C to 330 deg.C, Molecular Weight: 177.163, Percent Purity: 98%, Beilstein: 383929, CAS: 521-31-3, EINECS Number: 208-309-4, Formula Weight: 177.16, Health Hazard 1: H315-H319-H335

3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00006890
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

Melting Point 325°C to 330°C
Odor Odorless
Quantity 5 g
Beilstein 383929
Merck Index 14,5600
Solubility Information Soluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 3-Aminophthalhydrazide
compliance-icons
Product Identifier
  • 3-Aminophthalhydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 5-amino-2,3-dihydro-1,4-Phthalazinedione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.