Learn More
3-Aminopropionitrile, 98%, stabilized, Thermo Scientific Chemicals
32.36 € - 329.64 €
Chemical Identifiers
CAS | 151-18-8 |
---|---|
Molecular Formula | C3H6N2 |
Molecular Weight (g/mol) | 70.10 |
MDL Number | MFCD00014820 |
InChI Key | AGSPXMVUFBBBMO-UHFFFAOYSA-N |
Synonym | 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino |
PubChem CID | 1647 |
ChEBI | CHEBI:27413 |
IUPAC Name | 3-aminopropanenitrile |
SMILES | NCCC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10684272
|
Thermo Scientific Acros
351750010 |
1 g | Glass bottle |
32.36 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10567021
|
Thermo Scientific
351750050 |
5 g | Glass bottle |
99.09 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925621
|
Thermo Scientific Acros
351750250 |
25 g | Glass bottle |
329.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151-18-8 | |
70.10 | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
1647 | |
3-aminopropanenitrile |
C3H6N2 | |
MFCD00014820 | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
CHEBI:27413 | |
NCCC#N |
Specifications
151-18-8 | |
100.0 | |
0.9584g/mL | |
85°C | |
97.5% min. (GC) | |
1.4365 to 1.4395 | |
15,465 | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
3-aminopropanenitrile | |
70.10 | |
CHEBI:27413 | |
98% | |
3-Aminopropionitrile, Stabilized |
97.5 | |
Colorless to Yellow | |
185.0°C | |
Authentic | |
C3H6N2 | |
MFCD00014820 | |
0.9584 | |
Solubility in water: miscible. | |
NCCC#N | |
potassium carbonate | |
1647 | |
70.09 | |
Liquid |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-786-
- 3-Aminopropionitrile, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H361-Suspected of damaging fertility or the unborn child.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.