Learn More
(3-Aminopropyl)triethoxysilane, 98%
Descripción
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.
Solubility
Miscible with toluene, acetone, chloroform and ethanol.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.
Especificaciones
Especificaciones
| Boiling Point | 217°C |
| pH | 11 |
| Quantity | 100 g |
| Linear Formula | H2N(CH2)3Si(OCH2CH3)3 |
| UN Number | UN2735 |
| Beilstein | 1754988 |
| Flash Point | 104°C (219°F) |
| Solubility Information | Miscible with toluene,acetone,chloroform and ethanol. |
| Formula Weight | 221.37 |
| Percent Purity | 98% |
| Mostrar más |
Seguridad y manipulación
- (3-Aminopropyl)triethoxysilane
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 3-(Triethoxysilyl) propylamine
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.