missing translation for 'onlineSavingsMsg'
Learn More
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals Product Code.: 11493644

(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11493644
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11493644

Brand: Thermo Scientific Alfa Aesar A11284.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.291
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6Show MoreShow Less
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

Density 1.02
Boiling Point 194°C
Flash Point 90°C (194°F)
Linear Formula H2N(CH2)3Si(OCH3)3
Refractive Index 1.424
Quantity 100 g
UN Number UN2735
Beilstein 2038118
Sensitivity Air and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 179.29
Percent Purity 97%
Chemical Name or Material (3-Aminopropyl)trimethoxysilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253837 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA11284.22,A11284.0E,A11284.36
Lot Number10249777 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog NumberA11284.22,A11284.0E,A11284.36
Lot Number10244146 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog NumberA11284.22,A11284.0E,A11284.36
Lot Number10239306 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11284.22,A11284.0E,A11284.36
Lot Number10156787 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11284.22,A11284.0E,A11284.36
compliance-icons
Product Identifier
  • (3-Aminopropyl)trimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : (3-Aminopropyl)trimethoxysilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(3-Aminopropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.