Learn More
3-Aminopyridazine, 97%, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Spécification
Spécification
Melting Point | 168°C to 172°C |
Color | White to Yellow |
Quantity | 1 g |
Beilstein | 107024 |
Formula Weight | 95.1 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 3-Aminopyridazine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N27E035 | Certificate of Analysis | 11/07/2023 | H64859,H64859.06,H64859.03,H64859.14 |
Z19I048 | Certificate of Analysis | 11/01/2023 | H64859.06,H64859.03,H64859.14 |
81500409 | Certificate of Analysis | 08/12/2021 | H64859.06,H64859.03,H64859.14 |
81302855 | Certificate of Analysis | 06/12/2021 | H64859.06,H64859.03,H64859.14 |
N27D056 | Certificate of Analysis | 06/12/2021 | H64859.06,H64859.03,H64859.14 |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.