missing translation for 'onlineSavingsMsg'
Learn More
3-Aminopyridine, 99%, Thermo Scientific Chemicals Product Code.: 11410733

3-Aminopyridine, 99%, Thermo Scientific Chemicals

Product Code. 11410733
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11410733

Brand: Thermo Scientific Alfa Aesar A13216.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

Solubility
It is soluble in water as well as soluble in alcohol, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006400
InChI Key CUYKNJBYIJFRCU-UHFFFAOYSA-N
Synonym 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine
PubChem CID 10009
IUPAC Name pyridin-3-amine
SMILES NC1=CC=CN=C1

Specifications

Melting Point 58°C to 63°C
pH 10
Boiling Point 250°C to 252°C
Flash Point 88°C (190°F)
Odor Unpleasant
Quantity 1000 g
UN Number UN2671
Beilstein 105692
Sensitivity Hygroscopic
Merck Index 14,473
Solubility Information It is soluble in water as well as soluble in alcohol,benzene.
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 3-Aminopyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10221451 Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberA13216.30,A13216.18,A13216.0B
Lot Number10242577 Certificate TypeCertificate of Analysis Date09/08/2022 Catalog NumberA13216.30,A13216.18,A13216.0B
Lot Number10238250 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13216.30,A13216.18,A13216.0B
Lot Number10166818 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13216.30,A13216.18,A13216.0B
Lot Number10216994 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13216.30,A13216.18,A13216.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminopyridine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.