missing translation for 'onlineSavingsMsg'
Learn More
3-Aminotetrahydro-1H-1lambda∼6∼-thiophene-1,1-dione hydrochloride , Tech., Thermo Scientific™ Product Code.: 10095112

3-Aminotetrahydro-1H-1lambda∼6∼-thiophene-1,1-dione hydrochloride , Tech., Thermo Scientific™

Product Code. 10095112
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10095112

Brand: Thermo Scientific Maybridge RJC02399DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51642-03-6
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.64
MDL Number MFCD00456584
InChI Key MGZQMSFXPSKBDY-UHFFFAOYNA-N
Synonym 3-aminotetrahydrothiophene 1,1-dioxide hydrochloride, 1,1-dioxidotetrahydrothien-3-ylamine hydrochloride, tetrahydro-3-thiophenamine 1,1-dioxide hydrochloride, 1,1-dioxo-tetrahydrothiophen-3-ylamine, hcl, 3-amino-1??-thiolane-1,1-dione hydrochloride, 1,1-dioxidotetrahydro-3-thienyl amine hydrochloride, 3-aminotetrahydrothiopene 1,1-dioxide hydrochloride, tetrahydrothiophen-3-amine 1,1-dioxide hydrochloride, 3-thiophenamine, tetrahydro-, 1,1-dioxide, hydrochloride, 1,1-dioxothiolan-3-amine hydrochloride
PubChem CID 2795201
SMILES [H+].[Cl-].NC1CCS(=O)(=O)C1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 1,1-dioxothiolan-3-amine;hydrochloride
Formula Weight 171.65
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Aminotetrahydrothiopene 1, 1-dioxide hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Aminotetrahydro-1H-1lambda∼6∼-thiophene-1,1-dione hydrochloride , Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.