missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-5-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15449447
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-56-9
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.04
MDL Number MFCD07363782
InChI Key QQOFKMPPQQMUDH-UHFFFAOYSA-N
Synonym 3-benzyloxy-5-fluorophenylboronic acid, 3-benzyloxy-5-fluorobenzeneboronic acid, 3-benzyloxy-5-fluoro benzeneboronic acid, 3-phenylmethoxy-5-fluorobenzeneboronic acid, 3-benzyloxy-5-fluorophenyl boronic acid, boronic acid,b-3-fluoro-5-phenylmethoxy phenyl, b-3-fluoro-5-phenylmethoxy phenyl-boronic acid, boronic acid, b-3-fluoro-5-phenylmethoxy phenyl
PubChem CID 44558161
IUPAC Name (3-fluoro-5-phenylmethoxyphenyl)boronic acid
SMILES OB(O)C1=CC(OCC2=CC=CC=C2)=CC(F)=C1

Specifications

Melting Point 156°C to 162°C
Quantity 250 mg
Formula Weight 246.04
Percent Purity 98%
Chemical Name or Material 3-Benzyloxy-5-fluorobenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.