missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)phenol, 97%, Thermo Scientific Chemicals

Product Code. 15411188
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19962-06-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD06798070
InChI Key HJQNVUQTARSZDK-UHFFFAOYSA-N
Synonym tert-butyl 3-hydroxyphenylcarbamate, 3-boc-amino phenol, tert-butyl n-3-hydroxyphenyl carbamate, tert-butyl 3-hydroxyphenyl carbamate, n-boc-3-aminophenol, carbamic acid, 3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid,n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, tert-butoxy-n-3-hydroxyphenyl carboxamide, n-boc-3-amino-phenol
PubChem CID 312485
IUPAC Name tert-butyl N-(3-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC(O)=C1

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)phenol
compliance-icons
Product Identifier
  • 3-(Boc-amino)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-(Boc-amino)phenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.