Learn More
(+/-)-3-(Boc-amino)pyrrolidine, 97%, Thermo Scientific Chemicals
Description
(±)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(+/-)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.
Solubility
Soluble in Methanol(almost transparency).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Specifications
Specifications
Melting Point | 54°C |
Solubility Information | Soluble in Methanol(almost transparency). |
Quantity | 5 g |
Formula Weight | 186.25 |
Sensitivity | Air Sensitive |
Percent Purity | 97% |
Chemical Name or Material | (+/-)-3-(Boc-amino)pyrrolidine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
K02W071 | Certificate of Analysis | 07/12/2021 | H52414.03,H52414.06 |
S03G007 | Certificate of Analysis | 07/12/2021 | H52414.03,H52414.06 |
T23B027 | Certificate of Analysis | 07/12/2021 | H52414.03,H52414.06 |
L05X015 | Certificate of Analysis | 07/12/2021 | H52414.03,H52414.06 |
W22H049 | Certificate of Analysis | 07/12/2021 | H52414.03,H52414.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.