missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-aminomethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15459197
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199609-62-6
Molecular Formula C12H18BNO4
Molecular Weight (g/mol) 251.089
MDL Number MFCD06246052
InChI Key YHAQUGOSDQZIMA-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethyl phenylboronic acid, 3-tert-butoxycarbonyl amino methyl phenyl boronic acid, 3-n-boc-amino methyl phenylboronic acid, 3-n-boc aminomethylphenylboronic acid, 3-tert-butoxycarbonylamino methyl phenylboronic acid, 3-n-boc-aminomethyl phenyl boronic acid, 3-tert-butoxycarbonylaminomethyl phenylboronic acid, 3-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 3-aminomethyl benzeneboronic acid, n-boc protected, 3-tert-butoxycarbonyl amino methyl phenylboronic acid
PubChem CID 3684686
IUPAC Name [3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)CNC(=O)OC(C)(C)C)(O)O

Specifications

Melting Point 188°C to 190°C
Quantity 1 g
Formula Weight 251.09
Percent Purity 97%
Chemical Name or Material 3-(Boc-aminomethyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'