missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-cyanobenzeneboronic acid, 96%, Thermo Scientific Chemicals

47.50 € - 153.00 €

Chemical Identifiers

CAS 1032231-32-5
Molecular Formula C7H5BBrNO2
Molecular Weight (g/mol) 225.836
InChI Key LHGIIQFOQAPPQA-UHFFFAOYSA-N
Synonym 3-bromo-2-cyanophenyl boronic acid, 2-cyano-3-bromophenylboronic acid
PubChem CID 46739320
IUPAC Name (3-bromo-2-cyanophenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)Br)C#N)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418468
View Documents
Thermo Scientific Alfa Aesar
H61871.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15408468
View Documents
Thermo Scientific Alfa Aesar
H61871.03
1 g
153.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1032231-32-5
225.836
3-bromo-2-cyanophenyl boronic acid, 2-cyano-3-bromophenylboronic acid
(3-bromo-2-cyanophenyl)boronic acid
C7H5BBrNO2
LHGIIQFOQAPPQA-UHFFFAOYSA-N
46739320
B(C1=C(C(=CC=C1)Br)C#N)(O)O

Specifications

1032231-32-5
3-bromo-2-cyanophenyl boronic acid, 2-cyano-3-bromophenylboronic acid
B(C1=C(C(=CC=C1)Br)C#N)(O)O
225.836
225.84
Solid
C7H5BBrNO2
LHGIIQFOQAPPQA-UHFFFAOYSA-N
(3-bromo-2-cyanophenyl)boronic acid
46739320
96%
3-Bromo-2-cyanobenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.