Learn More
3-Bromo-2-methylpropene, 97%, Thermo Scientific Chemicals
67.74 € - 223.11 €
Chemical Identifiers
CAS | 1458-98-6 |
---|---|
Molecular Formula | C4H7Br |
Molecular Weight (g/mol) | 135.00 |
MDL Number | MFCD00134155 |
InChI Key | USEGQJLHQSTGHW-UHFFFAOYSA-N |
Synonym | 3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide |
PubChem CID | 357785 |
IUPAC Name | 3-bromo-2-methylprop-1-ene |
SMILES | CC(=C)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10149599
|
Thermo Scientific Acros
427650050 |
5 mL | Glass bottle |
67.74 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10832251
|
Thermo Scientific Acros
427650250 |
25 mL | Glass bottle |
223.11 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1458-98-6 | |
135.00 | |
USEGQJLHQSTGHW-UHFFFAOYSA-N | |
357785 | |
CC(=C)CBr |
C4H7Br | |
MFCD00134155 | |
3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide | |
3-bromo-2-methylprop-1-ene |
Specifications
1458-98-6 | |
100.0 | |
94.0°C to 95.0°C | |
96% min. (GC) | |
CH2=C(CH3)CH2Br | |
1.339 | |
USEGQJLHQSTGHW-UHFFFAOYSA-N | |
3-bromo-2-methylprop-1-ene | |
357785 | |
97% |
96.0 | |
1.3390g/mL | |
7°C | |
C4H7Br | |
MFCD00134155 | |
3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide | |
CC(=C)CBr | |
135.00 | |
135 | |
3-Bromo-2-methylpropene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away fr
GHS Signal Word: Danger
- 3-Bromo-2-methylpropene
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.