missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 15417678
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7138-15-0
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD08063248
InChI Key GLKHLBAXHLAQBJ-UHFFFAOYSA-N
Synonym 3-bromo-2-nitrobenzenamine, 2-amino-6-bromonitrobenzene, 3-bromo-2-nitrophenylamine, 3-bromo-2-nitro-phenylamine, pubchem3139, acmc-209oig, 3-bromo-2-nitro-aniline, ksc376s4f, 3-amino-2-nitrobromobenzene
PubChem CID 21852072
IUPAC Name 3-bromo-2-nitroaniline
SMILES C1=CC(=C(C(=C1)Br)[N+](=O)[O-])N

Specifications

Color Red
Quantity 5 g
UN Number UN2811
Formula Weight 217.02
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 3-Bromo-2-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'