missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-cyanopyridine, 95%, Thermo Scientific™

Product Code. 11436090
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13958-98-0
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234143
InChI Key VPDIXJCBBUZNPO-UHFFFAOYSA-N
Synonym 3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo
PubChem CID 11074051
IUPAC Name 3-bromopyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)Br

Specifications

Melting Point 92°C to 94°C
Odor Odorless
Quantity 1 g
Formula Weight 183.01
Percent Purity 95%
Chemical Name or Material 3-Bromo-4-cyanopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'