missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-cyanopyridine, 97%, Thermo Scientific™

Product Code. 15239766
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 13958-98-0
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
InChI Key VPDIXJCBBUZNPO-UHFFFAOYSA-N
PubChem CID 11074051
IUPAC Name 3-bromopyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)Br

Specifications

Melting Point 91°C to 94°C
Assay Percent Range 96% min. (GC)
Quantity 1 g
Synonym 3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-cyanopyridine
compliance-icons
Product Identifier
  • 3-Bromo-4-cyanopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'