missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluoro-1,1'-biphenyl, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 251.1 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-4-fluoro-1, 1'-biphenyl |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'