missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluoroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Specifications
Melting Point | 34°C to 36°C |
Boiling Point | 235°C |
Quantity | 5 g |
UN Number | UN2811 |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 190.01 |
Percent Purity | 98% |
Chemical Name or Material | 3-Bromo-4-fluoroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'