missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzoic acid, 96%, Thermo Scientific Chemicals

Product Code. 11470598
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1007-16-5
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00042463
InChI Key ONELILMJNOWXSA-UHFFFAOYSA-M
Synonym 3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid
PubChem CID 70509
SMILES [O-]C(=O)C1=CC=C(F)C(Br)=C1

Specifications

Melting Point 138°C to 141°C
Quantity 1 g
IUPAC Name 3-bromo-4-fluorobenzoic acid
Formula Weight 219.01
Percent Purity 96%
Chemical Name or Material 3-Bromo-4-fluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'