missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11420513
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-58-1
Molecular Formula C8H6BrO3
Molecular Weight (g/mol) 230.04
MDL Number MFCD00020295
InChI Key BBPZABXVRBFWGD-UHFFFAOYSA-M
Synonym 3-bromo-p-anisic acid, benzoic acid, 3-bromo-4-methoxy, 3-bromo-4-methoxybenzoicacid, akos bbb/210, rarechem al bo 0751, timtec-bb sbb009955, 4-methoxy-3-bromo benzoic acid, 3-bromanissaure, pubchem2675, 2-bromo-4-carboxyanisole
PubChem CID 66836
IUPAC Name 3-bromo-4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1Br)C([O-])=O

Specifications

Melting Point 217°C to 222°C
Odor Odorless
Quantity 25 g
Beilstein 2087585
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'