missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methoxypyridine, 97%, Thermo Scientific™

Product Code. 10386724
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50720-12-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00234169
InChI Key FZWUIWQMJFAWJW-UHFFFAOYSA-N
Synonym 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
PubChem CID 817163
IUPAC Name 3-bromo-5-methoxypyridine
SMILES COC1=CC(=CN=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 31°C to 35°C
Color White to Yellow
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3-Bromo-5-methoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-5-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.