missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-5-(trifluoromethyl)aniline, 98%, Thermo Scientific™ Product Code.: 11404397

3-Bromo-5-(trifluoromethyl)aniline, 98%, Thermo Scientific™

Product Code. 11404397
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11404397

Brand: Thermo Scientific Alfa Aesar B20437.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54962-75-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
MDL Number MFCD00236205
InChI Key HJTLKVYOWNTDPF-UHFFFAOYSA-N
Synonym 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
PubChem CID 2735880
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)Br)C(F)(F)F

Specifications

Density 1.71
Boiling Point 221°C to 223°C
Flash Point 107°C (225°F)
Refractive Index 1.5286
Quantity 1 g
UN Number UN2810
Formula Weight 240.03
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10213293 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10162329 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10192220 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberH4534A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10216127 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-5-(trifluoromethyl)aniline, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.